Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1-fluoro-4-(trifluoromethoxy)benzene 95%

Supplier: APOLLO SCIENTIFIC

PC910138-1G PC910138-250MG PC910138-5G
77418-566EA 371.86 USD InStock
77418-566 77418-568 77418-570
2-Chloro-1-fluoro-4-(trifluoromethoxy)benzene 95%
2-Chloro-1-fluoro-4-(trifluoromethoxy)benzene


Formula: C₇H₃ClF₄O
Storage Temperature: Ambient
MDL Number: MFCD22580940
CAS Number: 128540-52-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty