Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-(triphenylphosphoranylidene)acetate 94%

Supplier: APOLLO SCIENTIFIC
Warning

OR10645-100G OR10645-25G OR10645-500G
77402-858EA 28.52 USD InStock
77402-858 77402-860 77402-862
Ethyl-2-(triphenylphosphoranylidene)acetate 94%
Ethyl-2-(triphenylphosphoranylidene)acetate


Formula: C₂₂H₂₁O₂P
Melting Pt: 128…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009183
CAS Number: 1099-45-2
EINECS: 214-151-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty