Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR1170T-1G OR1170T-250MG OR1170T-5G
77404-026EA 103.92 USD InStock
77404-026 77404-028 77404-030
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside 97%
5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside


Formula: C₉H₁₄N₄O₅
Melting Pt: 214…215 °C
Storage Temperature: Freezer
MDL Number: MFCD00869751
CAS Number: 2627-69-2
EINECS: 220-097-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty