Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR29355-1G OR29355-250MG OR29355-5G
77408-384EA 303.6 USD InStock
77408-384 77408-386 77408-388
3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole
3-(Chloromethyl)-5-cyclobutyl-1,2,4-oxadiazole


Formula: C₇H₉ClN₂O
MW: 172.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00728735
CAS Number: 259683-88-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty