Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl dibromomalonate

Supplier: APOLLO SCIENTIFIC
Danger

OR46029-100G OR46029-25G
77409-878EA 84.84 USD InStock
77409-878 77409-880 77409-882
Diethyl dibromomalonate
Diethyl dibromomalonate


Formula: C₇H₁₀Br₂O₄
MW: 317.96 g/mol
Boiling Pt: 108…110 °C/4 mmHg
Density: 1.68 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015154
CAS Number: 631-22-1
EINECS: 211-151-9
UN: 1760
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty