Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl cyano(hydroxyimino)acetate

Supplier: APOLLO SCIENTIFIC
Warning

OR29185-100G OR29185-250G OR29185-25G
77408-358EA 41.78 USD InStock
77408-358 77408-360 77408-362
Ethyl cyano(hydroxyimino)acetate
Ethyl cyano(hydroxyimino)acetate


Formula: C₅H₆N₂O₃
MW: 142.11 g/mol
Melting Pt: 127…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00002112
CAS Number: 3849-21-6
EINECS: 223-351-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty