Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Ethoxycarbonyl)phenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR10468-25G OR10468-5G
77402-726EA 79.46 USD InStock
77402-726 77402-728
2-(Ethoxycarbonyl)phenylboronic acid 97%
2-(Ethoxycarbonyl)phenylboronic acid


Formula: C₉H₁₁BO₄
MW: 193.99 g/mol
Melting Pt: 128…136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179453
CAS Number: 380430-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty