Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate 97%

Supplier: APOLLO SCIENTIFIC

OR13735-10G OR13735-25G OR13735-5G
77404-728EA 172.18 USD InStock
77404-728 77404-730 77404-732
Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate 97%
Ethyl 5-amino-1,3,4-thiadiazole-2-carboxylate


Formula: C₅H₇N₃O₂S
MW: 173.193 g/mol
Melting Pt: 197…199 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093787
CAS Number: 64837-53-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty