Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodouracil 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR18306-100G OR18306-25G OR18306-500G
77406-218EA 75.4 USD InStock
77406-218 77406-220 77406-222
5-Iodouracil 97%
5-Iodouracil


Formula: C₄H₃IN₂O₂
Melting Pt: 274…276 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006020
CAS Number: 696-07-1
EINECS: 211-788-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty