Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gallium(III) trifluoromethanesulfonate

Supplier: APOLLO SCIENTIFIC
Danger

PC1246-10G PC1246-25G PC1246-5G
77413-106EA 131.42 USD InStock
77413-106 77413-108 77413-110
Gallium(III) trifluoromethanesulfonate
Gallium(III) trifluoromethanesulfonate


Formula: C₃F₉GaO₉S₃
Density: 1.7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02093898
CAS Number: 74974-60-0
UN: 3260
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty