Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pentenoic acid 98%

Supplier: APOLLO SCIENTIFIC

OR912162-100G OR912162-25G
77411-654EA 52.98 USD InStock
77411-654 77411-656
4-Pentenoic acid 98%
4-Pentenoic acid


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 187
Melting Pt: -18
Storage Temperature: Ambient
MDL Number: MFCD00004408
CAS Number: 591-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty