Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyethanesulfonic acid 80% in water 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR965087-100G OR965087-500G
77412-262EA 96.78 USD InStock
77412-262 77412-264
2-Hydroxyethanesulfonic acid 80% in water 97%
2-Hydroxyethanesulfonic acid


Formula: C₂H₆O₄S
Storage Temperature: Ambient
MDL Number: MFCD00242599
CAS Number: 107-36-8
UN: 3265
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty