Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-((Difluoromethyl)thio)benzoyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC0276-1G PC0276-5G
77412-478EA 151.8 USD InStock
77412-478 77412-480
3-((Difluoromethyl)thio)benzoyl chloride
3-((Difluoromethyl)thio)benzoyl chloride


Formula: C₈H₅ClF₂OS
Boiling Pt: 85…88 °C/8 mmHg
Storage Temperature: Ambient
CAS Number: 261944-16-5
UN: 1760
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty