Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-4-fluorobutane 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC3725-1G PC3725-25G PC3725-5G
77414-844EA 28.52 USD InStock
77414-844 77414-846 77414-848
1-Bromo-4-fluorobutane 97%
1-Bromo-4-fluorobutane


Formula: BrH₂C(CH₂)₂CH₂F
Boiling Pt: 134…135 °C
Density: 1.444 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00209551
CAS Number: 462-72-6
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty