Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloropropene 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR5736-1KG OR5736-500G OR5736-100G
77410-534EA 385.12 USD InStock
77410-534 77410-536 77410-532
2,3-Dichloropropene 97%
2,3-Dichloropropene


Formula: C₃H₄Cl₂
MW: 110.97 g/mol
Boiling Pt: 92.5
Melting Pt: -83.1
Density: 1.209 g/cm³ (25 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000943
CAS Number: 78-88-6
UN: 2047
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty