Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Hexyne

Supplier: APOLLO SCIENTIFIC
Danger

OR0049-250G OR0049-50G
77401-658EA 380.02 USD InStock
77401-658 77401-660
1-Hexyne
1-Hexyne


Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 71
Melting Pt: -132
Density: 0.715 g/cm³ (25 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00009504
CAS Number: 693-02-7
EINECS: 211-736-9
UN: 3295
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty