Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dibromobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR13221-25G OR13221-100G OR13221-250G
77404-594EA 21.36 USD InStock
77404-594 77404-590 77404-592
1,2-Dibromobenzene 98%
1,2-Dibromobenzene


Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 224
Melting Pt: 4…6 °C
Density: 1.956 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00000057
CAS Number: 583-53-9
EINECS: 209-507-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty