Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dibromo-4,5-difluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC2231-25G PC2231-5G
77413-566EA 23.44 USD InStock
77413-566 77413-568
1,2-Dibromo-4,5-difluorobenzene 98%
1,2-Dibromo-4,5-difluorobenzene
see: J. Chem. Soc., Perkin 1, 2729 (1995), a study of lithiation under different conditions.


Formula: C₆H₂Br₂F₂
Melting Pt: 34 °C
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00009890
CAS Number: 64695-78-9
EINECS: 265-021-1
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty