Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromoaniline 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR0036-1G OR0036-25G OR0036-5G
77401-648EA 21.4 USD InStock
77401-648 77401-650 77401-652
2,5-Dibromoaniline 95%
2,5-Dibromoaniline


Formula: C₆H₅Br₂N
Melting Pt: 51…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007636
CAS Number: 3638-73-1
EINECS: 222-865-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty