Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5397-100G OR5397-25G OR5397-500G
77410-344EA 55.02 USD InStock
77410-344 77410-346 77410-348
2-Bromophenol 98%
2-Bromophenol


Formula: C₆H₅BrO
MW: 173.01
Boiling Pt: 195
Melting Pt: 5.6
Density: 1.49 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002146
CAS Number: 95-56-7
EINECS: 202-432-7
UN: 1993
ADR: 3 III
Merck Index: 13,01413
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty