Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxysalicylaldehyde 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR1949-100G OR1949-25G OR1949-5G
77406-464EA 565.44 USD InStock
77406-464 77406-466 77406-468
5-Methoxysalicylaldehyde 98%
5-Methoxysalicylaldehyde


Formula: C₈H₈O₃
Boiling Pt: 103 °C/2.5 mmHg
Melting Pt: 4 °C
Density: 1.219 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty