Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichlorobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR0935-100G OR0935-25G
77402-316EA 23.34 USD InStock
77402-316 77402-318
3,5-Dichlorobenzoic acid
3,5-Dichlorobenzoic acid


Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 309
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00002494
CAS Number: 51-36-5
EINECS: 200-092-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty