Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloro-3-fluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1850-100G PC1850-25G PC1850-500G
77413-342EA 73.36 USD InStock
77413-342 77413-344 77413-346
1-Chloro-3-fluorobenzene 98%
1-Chloro-3-fluorobenzene


Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 126
Density: 1.219 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000569
CAS Number: 625-98-9
EINECS: 210-919-0
UN: 1993
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty