Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dichlorophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0058-100G OR0058-250G
77401-664EA 135.5 USD InStock
77401-664 77401-666
3,4-Dichlorophenol 98%
3,4-Dichlorophenol


Formula: C₆H₄Cl₂O
MW: 163
Boiling Pt: 247
Melting Pt: 64…68 °C
Density: 1.6
Storage Temperature: Ambient
MDL Number: MFCD00002258
CAS Number: 95-77-2
EINECS: 202-450-5
UN: 2020
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty