Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Difluorobenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2683-100G PC2683-25G PC2683-500G
77413-752EA 36.68 USD InStock
77413-752 77413-754 77413-756
3,4-Difluorobenzonitrile 98%
3,4-Difluorobenzonitrile


Formula: C₇H₃F₂N
Boiling Pt: 180 °C
Melting Pt: 48…54 °C
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00011666
CAS Number: 64248-62-0
EINECS: 264-751-8
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty