Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3400-100G PC3400-25G PC3400-500G
77414-714EA 135.5 USD InStock
77414-714 77414-716 77414-718
3-Fluoroaniline 98%
3-Fluoroaniline


Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 186
Melting Pt: -2
Density: 1.156 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007758
CAS Number: 372-19-0
EINECS: 206-747-0
UN: 2941
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty