Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3-Trifluoro-5-nitrobenzene

Supplier: APOLLO SCIENTIFIC
Danger

PC2435-25G PC2435-5G
77413-640EA 41.78 USD InStock
77413-640 77413-642
1,2,3-Trifluoro-5-nitrobenzene
1,2,3-Trifluoro-5-nitrobenzene


Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Density: 1.517 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00456803
CAS Number: 66684-58-0
EINECS: 266-447-0
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty