Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoroanisole 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC2614J-100G PC2614J-25G PC2614J-5G
77413-722EA 136.52 USD InStock
77413-722 77413-724 77413-726
3,5-Difluoroanisole 98%
3,5-Difluoroanisole


Formula: C₇H₆F₂O
Boiling Pt: 145…148 °C
Density: 1.234 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00042560
CAS Number: 93343-10-3
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty