Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-fluorotoluene

Supplier: APOLLO SCIENTIFIC
Warning

PC1900-100G PC1900-25G PC1900-5G
77413-404EA 65.54 USD InStock
77413-404 77413-406 77413-408
2-Chloro-5-fluorotoluene
2-Chloro-5-fluorotoluene


Formula: C₇H₆ClF
MW: 144.58
Boiling Pt: 156
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00040853
CAS Number: 33406-96-1
EINECS: 251-508-6
UN: 1993
ADR: 3 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty