Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1810-25G PC1810-100G
77413-314EA 45.26 USD InStock
77413-314 77413-312
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene 97%
4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene


Formula: C₇H₂ClF₃N₂O₄
MW: 270.55 g/mol
Boiling Pt: 250
Melting Pt: 50…58 °C
Density: 1.6 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007068
CAS Number: 393-75-9
EINECS: 206-889-3
UN: 2811
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty