Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-p-xylene 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR40462-100G OR40462-25G OR40462-500G
77409-542EA 150.78 USD InStock
77409-542 77409-544 77409-546
α-Bromo-p-xylene 98%
α-Bromo-p-xylene


Formula: C₈H₉Br
MW: 185.06
Boiling Pt: 218
Melting Pt: 38
Density: 1.324
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000181
CAS Number: 104-81-4
EINECS: 203-240-6
UN: 3417
ADR: 6.1 II
Merck Index: 12,10223
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty