Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Quinolinol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR301114-10G OR301114-1G OR301114-5G
77408-780EA 116.14 USD InStock
77408-780 77408-782 77408-784
7-Quinolinol 98%
7-Quinolinol


Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313
Melting Pt: 239…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00016730
CAS Number: 580-20-1
EINECS: 209-457-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty