Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluorocyclobutane 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC5920-100G PC5920-25G
77417-182EA 536 USD InStock
77417-182 77417-184 77417-186
Perfluorocyclobutane 99%
Perfluorocyclobutane


Formula: C₄F₈
Storage Temperature: Ambient
CAS Number: 115-25-3
EINECS: 204-075-2
UN: 1976
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty