Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromoacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR13196-100G OR13196-25G
77404-584EA 81.72 USD InStock
77404-584 77404-586
ω-Bromoacetophenone 97%
ω-Bromoacetophenone


Formula: (C₆H₅)C(O)CH₂Br
Melting Pt: 48…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000195
CAS Number: 70-11-1
EINECS: 200-724-9
UN: 2645
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty