Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Biphenylcarbonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR4021-250G OR4021-100G
77409-514EA 510.44 USD InStock
77409-514 77409-510 77409-512
4-Biphenylcarbonyl chloride 98%
4-Biphenylcarbonyl chloride


Formula: C₁₃H₉ClO
MW: 216.67 g/mol
Boiling Pt: 160 °C/2 mmHg
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00000692
CAS Number: 14002-51-8
EINECS: 237-804-8
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty