Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Perfluoro-tert-butanol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC5902-100G PC5902-25G PC5902-5G
77417-150EA 398.1 USD InStock
77417-150 77417-152 77417-154
Perfluoro-tert-butanol 97%
Perfluoro-tert-butanol


Formula: C₄HF₉O
MW: 236.04 g/mol
Boiling Pt: 84
Melting Pt: -17
Density: 1.693 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042092
CAS Number: 2378-02-1
EINECS: 219-157-3
UN: 2810
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty