Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Trifluoromethyl)thiophenol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7771-100G PC7771-25G PC7771-5G
77418-184EA 496.16 USD InStock
77418-184 77418-186 77418-188
2-(Trifluoromethyl)thiophenol 97%
2-(Trifluoromethyl)thiophenol


Formula: C₇H₅F₃S
Boiling Pt: 175…177 °C
Density: 1.35 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068896
CAS Number: 13333-97-6
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty