Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-methylpyridine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR5258-100G OR5258-25G OR5258-500G
77410-256EA 30.56 USD InStock
77410-256 77410-258 77410-260
2-Amino-3-methylpyridine 97%
2-Amino-3-methylpyridine


Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 221.1
Melting Pt: 29…32 °C
Density: 1.073 g/cm³ (25 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty