Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitro-2,3-xylidine

Supplier: APOLLO SCIENTIFIC
Warning

OR28632-25G OR28632-500G
77408-144EA 85.58 USD InStock
77408-144 77408-146
6-Nitro-2,3-xylidine
6-Nitro-2,3-xylidine


Formula: C₈H₁₀N₂O₂
Melting Pt: 115…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00010148
CAS Number: 59146-96-2
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty