Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium hydroxide 94+%

Supplier: APOLLO SCIENTIFIC
Danger

OR923866-50G OR923866-250G
77411-874EA 21.36 USD InStock
77411-874 77411-872
Barium hydroxide 94+%
Barium hydroxide


Formula: H₂BaO₂
MW: 171.34
Melting Pt: 325
Density: 2.18
Storage Temperature: Ambient
MDL Number: MFCD00003443
CAS Number: 17194-00-2
UN: 3262
ADR: 8 II
Merck Index: 12,01006
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty