Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium sulphide 75%, Technical Grade

Supplier: APOLLO SCIENTIFIC

IN1219-100G IN1219-25G
77401-360EA 131.42 USD InStock
77401-360 77401-362
Barium sulphide 75%, Technical Grade
Barium sulphide


Formula: BaS
MW: 169.4 g/mol
Melting Pt: 1999.85
Density: 4.25 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014192
CAS Number: 21109-95-5
Merck Index: 12,01024
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty