Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-2-Chloropropionic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR28944-100G OR28944-25G OR28944-5G
77408-294EA 111.06 USD InStock
77408-294 77408-296 77408-298
(S)-(-)-2-Chloropropionic acid
(S)-(-)-2-Chloropropionic acid


Formula: H₃CCHClCOOH
MW: 108.52 g/mol
Boiling Pt: 186
Melting Pt: 5
Density: 1.249 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00064205
CAS Number: 29617-66-1
EINECS: 411-150-2
UN: 2511
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty