Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorotoluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC6523-100G PC6523-25G PC6523-5G
77417-422EA 79.08 USD InStock
77417-422 77417-424 77417-426
2,3-Difluorotoluene 97%
2,3-Difluorotoluene


Formula: C₇H₆F₂
Boiling Pt: 119…121 °C
Density: 1.096 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD01631506
CAS Number: 3828-49-7
UN: 1993
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty