Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR49010-5G OR49010-100G OR49010-25G
77410-074EA 21.26 USD InStock
77410-074 77410-070 77410-072
4-Bromobenzylamine 98%
4-Bromobenzylamine


Formula: C₇H₈BrN
Boiling Pt: 110…112 °C/30 mmHg
Melting Pt: 25 °C
Density: 1.473 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00047931
CAS Number: 3959-07-7
EINECS: 223-559-4
UN: 3259
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty