Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4-Trifluorobenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7285N-1G PC7285N-25G PC7285N-5G
77417-728EA 21.26 USD InStock
77417-728 77417-730 77417-732
2,3,4-Trifluorobenzyl bromide 97%
2,3,4-Trifluorobenzyl bromide


Formula: C₇H₄BrF₃
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00061233
CAS Number: 157911-55-2
UN: 3265
ADR: 8,III
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty