Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Bromo-3,4-dichlorotoluene 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR21377-25G OR21377-5G
77406-678EA 74.38 USD InStock
77406-678 77406-680
α-Bromo-3,4-dichlorotoluene 97%
α-Bromo-3,4-dichlorotoluene


Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Boiling Pt: 115…120 °C/1 mmHg
Density: 1.65 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041363
CAS Number: 18880-04-1
UN: 3265
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty