Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-p-toluidine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC3818-100G PC3818-25G
77414-918EA 74.38 USD InStock
77414-918 77414-920
3-Fluoro-p-toluidine 98%
3-Fluoro-p-toluidine


Formula: C₇H₈FN
MW: 125.15
Boiling Pt: 97
Melting Pt: 37
Density: 1.093 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00007762
CAS Number: 452-77-7
EINECS: 207-212-4
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty