Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromophenyl isothiocyanate 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR3081-100G OR3081-25G OR3081-5G
77408-978EA 450.32 USD InStock
77408-978 77408-980 77408-982
3-Bromophenyl isothiocyanate 97%
3-Bromophenyl isothiocyanate


Formula: C₇H₄BrNS
MW: 214.09 g/mol
Boiling Pt: 256 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004803
CAS Number: 2131-59-1
UN: 3335
ADR: 9 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty