Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-α,α,α-trifluoro-5-nitrotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC1490-100G PC1490-25G
77413-198EA 50.94 USD InStock
77413-198 77413-200
2-Bromo-α,α,α-trifluoro-5-nitrotoluene 98%
2-Bromo-α,α,α-trifluoro-5-nitrotoluene


Formula: C₇H₃BrF₃NO₂
MW: 270.01
Boiling Pt: 87…88 °C/3 mmHg
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00014707
CAS Number: 367-67-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty