Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-fluoroanisole 95%

Supplier: APOLLO SCIENTIFIC

PC910089-1G PC910089-25G PC910089-5G
77418-560EA 29.54 USD InStock
77418-560 77418-562 77418-564
5-Chloro-2-fluoroanisole 95%
5-Chloro-2-fluoroanisole


Formula: C₇H₆ClFO
Storage Temperature: Ambient
CAS Number: 1092349-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty